missing translation for 'onlineSavingsMsg'
Learn More

Iminodiacetic acid, 98+%, Thermo Scientific Chemicals

Catalog Number 11494493
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
142-73-4
C4H7NO4
133.103
MFCD00004280
NBZBKCUXIYYUSX-UHFFFAOYSA-N
iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
8897
CHEBI:24786
2-(carboxymethylamino)acetic acid
C(C(=O)O)NCC(=O)O
This item is not returnable. View return policy
142-73-4
C4H7NO4
133.103
MFCD00004280
NBZBKCUXIYYUSX-UHFFFAOYSA-N
iminodiacetic acid, 2,2'-iminodiacetic acid, glycine, n-carboxymethyl, aminodiacetic acid, diglycocoll, diglykokoll, diglycin, iminodiethanoic acid, n-carboxymethyl glycine, 2,2'-azanediyldiacetic acid
8897
CHEBI:24786
2-(carboxymethylamino)acetic acid
C(C(=O)O)NCC(=O)O

Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼243°C (decomposition)
Quantity 100 g
Beilstein 878499
Merck Index 14,4917
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 133.1
Percent Purity ≥98%
Chemical Name or Material Iminodiacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'