missing translation for 'onlineSavingsMsg'
Learn More

Imidurea, 26-28% nitrogen, Thermo Scientific Chemicals

Catalog Number p-148112
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
39236-46-9
C11H16N8O8
388.29
MFCD00221482
ZCTXEAQXZGPWFG-UHFFFAOYSA-N
imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
38258
CHEBI:51805
1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO
This item is not returnable. View return policy
39236-46-9
C11H16N8O8
388.29
MFCD00221482
ZCTXEAQXZGPWFG-UHFFFAOYSA-N
imidazolidinyl urea, imidurea, germall 115, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxoimidazolidin-4-yl urea, imidazolinidyl urea, imidurea nf, methanebis n,n'-5-ureido-2,4-diketotetrahydroimidazole-n,n-dimethylol, n,n-methylenebis n'-1-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea, 1,1'-methylenebis 3-3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl urea
38258
CHEBI:51805
1-[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]-3-[[[3-(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl]carbamoylamino]methyl]urea
C(NC(=O)NC1C(=O)NC(=O)N1CO)NC(=O)NC2C(=O)NC(=O)N2CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
pH 6.0 to 7.5 (1% aq. soln.)
Loss on Drying 3.0 % max. (48 hrs) (vacuum, P2O5)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 4957
Solubility Information Solubility in water: soluble
Formula Weight 388.29
Percent Purity 26-28% nitrogen
Physical Form Liquid
Chemical Name or Material Imidurea, 26-28% nitrogen
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'