missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidocarb Dipropionate, TRC
Specifications
Specifications
| Chemical Name or Material | Imidocarb dipropionate |
| CAS | 55750-06-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H20 N6 O . 2 C3 H6 O2 |
| Quantity | 500 mg |
| Synonym | Imidocarb Dipropionate, Propanoic acid compd. with N,N'-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea (2:1), Carbesia, Imidocarb dipropionate, Imizol, Imizol (antiprotozoal), N,N'-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea dipropanoate |
| InChI Formula | InChI=1S/C19H20N6O.2C3H6O2/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18;2*1-2-3(4)5/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26);2*2H2,1H3,(H,4,5) |
| SMILES | CCC(=O)O.CCC(=O)O.O=C(Nc1cccc(c1)C2=NCCN2)Nc3cccc(c3)C4=NCCN4 |
| IUPAC Name | 1,3-bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea;propanoic acid |
| Molecular Weight (g/mol) | 496.56 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?