missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, MP Biomedicals™

Product Code. 11358859 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358859 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11358859 Supplier MP Biomedicals Supplier No. 02102033.1
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Excellent for buffer use in pH range 6.2–7.8. Imidazole, MP Biomedicals is a chelator for the binding of various divalent cations. It is a highly polar, planar 5 membered ring that is amphoteric in nature.

Imidazole can be used for the elution of histidine containing proteins from divalent cation resins and can also be used in reverse staining of SDS-PAGE gels for detection of proteins . Imidazole is used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column. Synthetic imidazoles are present in many fungicides and antifungal, antiprotozoal, and antihypertensive medications.

Specifications

CAS 288-32-4
Melting Point 86°C to 94°C
Density 500–600 kg/m3 (bulk density) (Lit.)
pH ≥7 (1% aq. soln.)
Boiling Point 268°C (literature)
Molecular Formula C3H4N2
MDL Number MFCD00005183
Quantity 100 g
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
Solubility Information Freely soluble in water, alcohol, ehter, chloroform, pyridine; slightly soluble in benzene; very sparingly soluble in petroleum ether.
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
SMILES N1C=CN=C1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.08
PubChem CID 795
ChEBI CHEBI:16069
Formula Weight 68.1
Vapor Pressure .003hPa at 20°C (Lit.)
Percent Purity ≥99%
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.