missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Molecular Biology Grade, 99+%, Thermo Scientific Chemicals

Product Code. p-7057670
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
This item is not returnable. View return policy

Product Code. 15418649

Brand: Thermo Scientific Alfa Aesar J67132.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
SMILES N1C=CN=C1

Specifications

Melting Point 90°C
Color White
Density 1.03g/mL
Boiling Point 256°C
Quantity 25 g
UN Number UN2923
Beilstein 103853
Merck Index 144,912
IUPAC Name 1H-imidazole
Formula Weight 68.08
Percent Purity ≥99%
Grade Molecular Biology
Physical Form Powder
Chemical Name or Material Imidazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.