missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, ACS reagent, Thermo Scientific Chemicals

Catalog Number 10557244
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
288-32-4
C3H4N2
68.08
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
795
CHEBI:16069
1H-imidazole
N1C=CN=C1
288-32-4
C3H4N2
68.08
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
795
CHEBI:16069
1H-imidazole
N1C=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 86.0°C to 90.0°C
Color White to Yellow
pH 9.5 to 11.0 (at 5% 25°C)
Boiling Point 255.0°C to 256.0°C
Flash Point 145°C
Ignition Residue 0.1% max.
Packaging Plastic bottle
Quantity 2.5 kg
Merck Index 15, 4953
Solubility Information Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether
Formula Weight 68.08
Percent Purity ≥99%
Grade ACS Reagent
Physical Form Crystals or Powder
Chemical Name or Material Imidazole, For analysis ACS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'