missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, 99+%, crystalline

Product Code. p-3736726 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10697821

Brand: Thermo Scientific Acros 301872500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Imidazole, 99+%, C3H4N2, CAS Number-288-32-4, glyoxalin, imidazole, miazole, imutex, pyrro b monazole, 1,3-diazole, imidazol, iminazole, 1,3-diaza-2,4-cyclopentadiene, glyoxaline, 1kg, 99% min. (GC), 23, 45, 255.0 deg.C to 256.0 deg.C, CHEBI:16069, White to Yellow or Beige, 206-019-2, 01,492; 02,220 | CAS: 288-32-4 | C3H4N2 | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-32-4
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005183
InChI Key RAXXELZNTBOGNW-UHFFFAOYSA-N
Synonym imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
PubChem CID 795
ChEBI CHEBI:16069
IUPAC Name 1H-imidazole
SMILES N1C=CN=C1

Specifications

Melting Point 86.0°C to 90.0°C
Color White to Yellow or Beige
Boiling Point 255.0°C to 256.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 250 g
Beilstein 23, 45
Fieser 01,492; 02,220
Merck Index 15, 4953
Solubility Information Solubility in water: 633g/L (20°C). Other solubilities: freely soluble in alcohol,ether,chloroform,slightly soluble in benzene,sparingly soluble in petroleum ether
Formula Weight 68.08
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Imidazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.