missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole, >99.6%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
:
5kg
Specifications
Specifications
| CAS | 288-32-4 |
| Color | White |
| Density | 500 - 600kg/m3 (bulk density) |
| Assay Percent Range | >99.6% |
| Molecular Formula | C3H4N2 |
| MDL Number | MFCD00005183 |
| Quantity | 5 kg |
| Synonym | imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole |
| InChI Key | RAXXELZNTBOGNW-UHFFFAOYSA-N |
| SMILES | N1C=CN=C1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction