missing translation for 'onlineSavingsMsg'
Learn More

Imidazole-4-carboxylic acid, 98%

Product Code. 15493408
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493408 1 g 1g
15413418 250 mg 250mg
15403418 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493408 Supplier Thermo Scientific Alfa Aesar Supplier No. H56862.03
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072-84-0 | C4H4N2O2 | 112.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-84-0
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.09
MDL Number MFCD00082203
InChI Key NKWCGTOZTHZDHB-UHFFFAOYSA-N
Synonym 1h-imidazole-4-carboxylic acid, 4-imidazolecarboxylic acid, imidazole-4-carboxylic acid, 3h-imidazole-4-carboxylic acid, 4-carboxyimidazole, unii-ae82z8u4gj, imidazole-5-carboxylic acid, 4-imidazole carboxylic acid, ae82z8u4gj, 4 5-imidazolecarboxylic acid
PubChem CID 14080
IUPAC Name 1H-imidazole-5-carboxylic acid
SMILES OC(=O)C1=CN=CN1

Specifications

Melting Point 294°C to 295°C
Quantity 1 g
Formula Weight 112.09
Percent Purity 98%
Chemical Name or Material Imidazole-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.