missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[2,1-b][1,3]thiazole-6-carboxylic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250mg
1g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1g |
Formula Weight | 168.18 |
Percent Purity | 97% |
Chemical Name or Material | Imidazo[2, 1-b][1, 3]thiazole-6-carboxylic acid |