missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carboxylic acid, 90%, Thermo Scientific™

Product Code. 10092254
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10092254 5 g 5g
10107842 250 mg 250mg
10331233 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092254 Supplier Thermo Scientific Maybridge Supplier No. CC33101DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 139022-25-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD07021498
InChI Key ONOJJCTXSDBVSP-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carboxylic acid, h-imidazo 1,2-a pyridine-6-carboxylic acid, imidazo 1,2-a pyridine-6-carboxylicacid, 1h-imidazo 1,2-a pyridine-6-carboxylic acid, 4-hydroimidazo 1,2-a pyridine-6-carboxylic acid, pubchem23163, acmc-209cig, ksc173s9l, 6-carboxyimidazo 1,2-a pyridine, imidazol 1,2-a pyridine-6-carboxylic acid
PubChem CID 7075376
IUPAC Name imidazo[1,2-a]pyridine-6-carboxylic acid
SMILES OC(=O)C1=CN2C=CN=C2C=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 162.15
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridine-6-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.