missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-2-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 15279796 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15279796 10 g 10g
15269796 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15279796 Supplier Thermo Scientific Acros Supplier No. 452640100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 118000-43-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
InChI Key QKKKBOZQZHBENG-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde, 2-formylpyridoimidazole, 2,3-dihydroimidazo 1,2-a pyridine-2-carbaldehyde, acmc-20ac55, imidazo 1,2-apyridine-2-carbaldehyde, imid-azo 1,2-a pyridine-2-carbaldehyde, 4-hydroimidazo 1,2-a pyridine-2-carbaldehyde, imidazo 1,2-a pyridine-2-carboxaldehyde 9ci
PubChem CID 4961257
IUPAC Name imidazo[1,2-a]pyridine-2-carbaldehyde
SMILES C1=CC2=NC(=CN2C=C1)C=O

Specifications

Melting Point 117.0°C to 119.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 10 g
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material Imidazo[1,2-a]pyridine-2-carboxaldehyde
compliance-icons
Product Identifier
  • Imidazo[1,2-a]pyridine-2-carboxaldehyde, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.