missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-6-ylmethanol, 97%, Thermo Scientific™

Product Code. 10401934
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10401934 5 g 5g
10269562 250 mg 250mg
10730154 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10401934 Supplier Thermo Scientific Maybridge Supplier No. CC33109DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 132213-07-1
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
InChI Key IEOADZOTWIMSMC-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-6-ylmethanol, 6-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-methanol, imidazo 1,2-a pyridin-6-yl-methanol, imidazo 1,2-a pyridin-6-yl methanol, acmc-209zmz, imidazo 1,2-a pyridin-6yl methanol, 6-hydroxymethyl imidazo 1,2-a pyridin, 4-hydroimidazo 1,2-a pyridin-6-ylmethan-1-ol
PubChem CID 7060530
IUPAC Name imidazo[1,2-a]pyridin-6-ylmethanol
SMILES C1=CC2=NC=CN2C=C1CO

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridin-6-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.