missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-2-ylmethanol, ≥90%, Thermo Scientific™

Product Code. 10289712
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10289712 1 g 1g
10719413 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10289712 Supplier Thermo Scientific Maybridge Supplier No. CC31309DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 82090-52-6
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.17
MDL Number MFCD06797471
InChI Key ROPHYEIJSUUKEO-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-2-ylmethanol, imidazo 1,2-a pyridine-2-methanol, h-imidazo 1,2-a pyridin-2-yl methanol, 2-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridin-2-yl methanol, imidazo 1,2-a-pyridin-2-ylmethanol, 4-hydroimidazo 1,2-a pyridin-2-ylmethan-1-ol, imidazo 1,2-a pyridine-2-ylmethanol
PubChem CID 2795540
SMILES OCC1=CN2C=CC=CC2=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name {imidazo[1,2-a]pyridin-2-yl}methanol
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridin-2-ylmethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.