missing translation for 'onlineSavingsMsg'
Learn More

Imidapril hydrochloride, MedChemExpress

Product Code. 16448766
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16448766

Brand: Medchem Express HYB1451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. Imidapril hydrochloride suppresses the conversion of angiotensin I to angiotensin II and thereby reduces total peripheral resistance and systemic blood pressure. Imidapril hydrochloride can be used for hypertension, type 1 diabetic, nephropathy and chronic heart failure research.

Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. Imidapril hydrochloride suppresses the conversion of angiotensin I to angiotensin II and thereby reduces total peripheral resistance and systemic blood pressure. Imidapril hydrochloride can be used for hypertension, type 1 diabetic, nephropathy and chronic heart failure research.

Specifications

Chemical Name or Material Imidapril hydrochloride
CAS 89396-94-1
Color White
Purity Grade Notes Research
Molecular Formula C20H28ClN3O6
Quantity 100 mg
Synonym TA-6366
Solubility Information DMSO : 50 mg/mL (113.15 mM; Need ultrasonic) ∣H2O : 50 mg/mL (113.15 mM; Need ultrasonic)
SMILES O=C([C@H](CN1C)N(C([C@@H](N[C@H](C(OCC)=O)CCC2=CC=CC=C2)C)=O)C1=O)O.[H]Cl
Molecular Weight (g/mol) 441.91
Formula Weight 441.91
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.