missing translation for 'onlineSavingsMsg'
Learn More

Imidapril hydrochloride, MedChemExpress

Product Code. 16494597
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494597 50 mg 50mg
16471456 200 mg 200mg
16448766 100 mg 100mg
16418708 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494597 Supplier Medchem Express Supplier No. HYB1451
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. Imidapril hydrochloride suppresses the conversion of angiotensin I to angiotensin II and thereby reduces total peripheral resistance and systemic blood pressure. Imidapril hydrochloride can be used for hypertension, type 1 diabetic, nephropathy and chronic heart failure research.

Imidapril hydrochloride (TA-6366) is an orally active angiotensin-converting enzyme (ACE) and MMP-9 inhibitor. Imidapril hydrochloride suppresses the conversion of angiotensin I to angiotensin II and thereby reduces total peripheral resistance and systemic blood pressure. Imidapril hydrochloride can be used for hypertension, type 1 diabetic, nephropathy and chronic heart failure research.

Specifications

Chemical Name or Material Imidapril hydrochloride
CAS 89396-94-1
Color White
Purity Grade Notes Research
Molecular Formula C20H28ClN3O6
Quantity 50 mg
Synonym TA-6366
Solubility Information DMSO : 50 mg/mL (113.15 mM; Need ultrasonic) ∣H2O : 50 mg/mL (113.15 mM; Need ultrasonic)
SMILES O=C([C@H](CN1C)N(C([C@@H](N[C@H](C(OCC)=O)CCC2=CC=CC=C2)C)=O)C1=O)O.[H]Cl
Molecular Weight (g/mol) 441.91
Formula Weight 441.91
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.