missing translation for 'onlineSavingsMsg'
Learn More

Imidacloprid-d4, TRC

Product Code. 30400269
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400269 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400269 Supplier Toronto Research Chemicals Supplier No. TRCI27499225MG
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imidacloprid D4 (imidazolidin-4,4,5,5 D4)
CAS 1015855-75-0
Purity Grade Notes HPLC
Molecular Formula C9 2H4 H6 Cl N5 O2
Quantity 25 mg
Synonym 1H-Imidazol-4,5-d2-2-amine, 1-[(6-chloro-3-pyridinyl)methyl]-4,5-dihydro-4,5-d2-N-nitro-, Imidacloprid D4 (imidazolidin-4,4,5,5 D4), Imidacloprid-d4
InChI Formula InChI=1S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5H,3-4,6H2,(H,11,13)/i3D2,4D2
SMILES [2H]C1([2H])N\C(=N/[N+](=O)[O-])\N(Cc2ccc(Cl)nc2)C1([2H])[2H]
IUPAC Name (NE)-N-[1-[(6-chloropyridin-3-yl)methyl]-4,4,5,5-tetradeuterioimidazolidin-2-ylidene]nitramide
Molecular Weight (g/mol) 259.69
Formula Weight 259.0774
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.