missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Imidacloprid
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 g |
Melting Point | 134.0°C to 146.0°C |
Formula Weight | 255.66 |
InChI Key | YWTYJOPNNQFBPC-UHFFFAOYSA-N |
SMILES | [O-][N+](=O)NC1=NCCN1CC1=CC=C(Cl)N=C1 |
IUPAC Name | 2-chloro-5-{[2-(nitroamino)-4,5-dihydro-1H-imidazol-1-yl]methyl}pyridine |
Molecular Weight (g/mol) | 255.66 |
Chemical Name or Material | Imidacloprid |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H301-Toxic if swallowed.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P405-Store locked up.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'