missing translation for 'onlineSavingsMsg'
Learn More

IMD 0354, TRC

Product Code. 30403086
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403086 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403086 Supplier Toronto Research Chemicals Supplier No. TRCI26865025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material N-(3,5-Bis(trifluoromethyl)phenyl)-5-chloro-2-hydroxy-benzamide
CAS 978-62-1
Molecular Formula C15H8ClF6NO2
Quantity 25 mg
Synonym IMD 0354, 5-Chloro-α,α,α,α',α',α'-hexafluoro-3',5'-salicyloxylidide N-(3,5-Bis(trifluoromethyl)phenyl)-5-chloro-2-hydroxybenzamide
InChI Formula InChI=1S/C15H8ClF6NO2/c16-9-1-2-12(24)11(6-9)13(25)23-10-4-7(14(17,18)19)3-8(5-10)15(20,21)22/h1-6,24H,(H,23,25)
SMILES Oc1ccc(Cl)cc1C(=O)Nc2cc(cc(c2)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 383.67
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.