missing translation for 'onlineSavingsMsg'
Learn More

Imazapic, MedChemExpress

Product Code. 16422296
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422296 500 mg 500mg
16418826 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422296 Supplier Medchem Express Supplier No. HYB1860
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Imazapic is a selective herbicide for both the preand post-emergent control of some annual and perennial grasses and some broadleaf weeds

Imazapic is a selective herbicide for both the preand post-emergent control of some annual and perennial grasses and some broadleaf weeds

Specifications

Chemical Name or Material Imazapic
CAS 104098-48-8
Color White
Purity Grade Notes Research
Molecular Formula C14H17N3O3
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (363.24 mM)
SMILES O=C(C1=CC(C)=CN=C1C2=NC(C(C)C)(C)C(N2)=O)O
Molecular Weight (g/mol) 275.3
Formula Weight 275.3
Percent Purity 99.86%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.