missing translation for 'onlineSavingsMsg'
Learn More

Imazamox, MedChemExpress

Product Code. p-200096518
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16493008

Brand: Medchem Express HY100427

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imazamox (CL29926) is a systemic herbicide that inhibits the production of acetolactate synthase (ALS) in plants with high selectivity, high activity, safety and broadspectrum activity, which would then inhibit plant growth and ultimately lead to plant death.

Imazamox (CL29926) is a systemic herbicide that inhibits the production of acetolactate synthase (ALS) in plants with high selectivity, high activity, safety and broadspectrum activity, which would then inhibit plant growth and ultimately lead to plant death.

Specifications

Chemical Name or Material Imazamox
CAS 114311-32-9
Color White
Purity Grade Notes Research
Molecular Formula C15H19N3O4
Quantity 10 mg
Synonym CL29926 (±)-Imazamox
Solubility Information DMSO : 20 mg/mL (65.50 mM; Need ultrasonic)
SMILES O=C(C1=CC(COC)=CN=C1C(NC2(C)C(C)C)=NC2=O)O
Molecular Weight (g/mol) 305.33
Formula Weight 305.33
Percent Purity 98.05%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.