missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imazalil Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Imazalil sulfate |
| CAS | 58594-72-2 |
| Molecular Formula | C14 H14 Cl2 N2 O . H2 O4 S |
| Quantity | 250 mg |
| Synonym | Imazalil Sulfate, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(2-propenyloxy)ethyl]-, sulfate (1:1) (9CI), Chloramizol sulfate, Imazalil sulfate, Microban Additive IF 4, R 27180, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(2-propen-1-yloxy)ethyl]-, sulfate (1:1), Enilconazole Sulfate |
| InChI Formula | InChI=1S/C14H14Cl2N2O.H2O4S/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16;1-5(2,3)4/h2-6,8,10,14H,1,7,9H2;(H2,1,2,3,4) |
| SMILES | OS(=O)(=O)O.Clc1ccc(C(Cn2ccnc2)OCC=C)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-prop-2-enoxyethyl]imidazole;sulfuric acid |
| Molecular Weight (g/mol) | 395.26 |
| Formula Weight | 394.0157 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?