missing translation for 'onlineSavingsMsg'
Learn More

Imatinib (Piperazine)-4-oxide, TRC

Product Code. 30397811
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397811 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397811 Supplier Toronto Research Chemicals Supplier No. TRCI26800010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imatinib (Piperidine)-N-oxide
CAS 571186-91-9
Purity Grade Notes HPLC
Molecular Formula C29 H31 N7 O2
Quantity 10 mg
Synonym Benzamide, 4-[(4-methyl-4-oxido-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-, 4-[(4-Methyl-4-oxido-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, CGP 71422, Imatinib (piperidine)-N-oxide
InChI Formula InChI=1S/C29H31N7O2/c1-21-5-10-25(18-27(21)34-29-31-13-11-26(33-29)24-4-3-12-30-19-24)32-28(37)23-8-6-22(7-9-23)20-35-14-16-36(2,38)17-15-35/h3-13,18-19H,14-17,20H2,1-2H3,(H,32,37)(H,31,33,34)
SMILES Cc1ccc(NC(=O)c2ccc(CN3CC[N+](C)([O-])CC3)cc2)cc1Nc4nccc(n4)c5cccnc5
IUPAC Name 4-[(4-methyl-4-oxidopiperazin-4-ium-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
Molecular Weight (g/mol) 509.6
Formula Weight 509.2539
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.