missing translation for 'onlineSavingsMsg'
Learn More

Imatinib Meta-methyl-piperazine Impurity, TRC

Product Code. 30388816
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388816 250 mg
30398801 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388816 Supplier Toronto Research Chemicals Supplier No. TRCI267990250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246819-59-9
Molecular Formula C29 H31 N7 O
Molecular Weight (g/mol) 493.6
Synonym 3-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, Benzamide, 3-[(4-methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]-
IUPAC Name 3-[(4-methylpiperazin-1-yl)methyl]-N-[4-methyl-3-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamide
SMILES CN1CCN(Cc2cccc(c2)C(=O)Nc3ccc(C)c(Nc4nccc(n4)c5cccnc5)c3)CC1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C29H31N7O/c1-21-8-9-25(18-27(21)34-29-31-12-10-26(33-29)24-7-4-11-30-19-24)32-28(37)23-6-3-5-22(17-23)20-36-15-13-35(2)14-16-36/h3-12,17-19H,13-16,20H2,1-2H3,(H,32,37)(H,31,33,34)
Formula Weight 493.259
Percent Purity >95%
Chemical Name or Material Imatinib meta-Methyl-piperazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.