missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imaradenant, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
AZD4635 (HTL1071) is a potent, selective and orally active adenosine A2A receptor (A2AR) antagonist. AZD4635 binds to human A2AR with a Ki of 1.7 nM and shows >30-fold selectivity over other adenosine receptors.
Specifications
Specifications
| Chemical Name or Material | Imaradenant |
| CAS | 1321514-06-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C15H11ClFN5 |
| Quantity | 10 mg |
| Synonym | HTL1071 AZD4635 |
| Solubility Information | DMSO : ≥ 83.3 mg/mL (263.83 mM) |
| SMILES | NC1=NC(C2=CC=C(F)C=C2)=C(C3=CC(Cl)=NC(C)=C3)N=N1 |
| Molecular Weight (g/mol) | 315.73 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction