Learn More
Motic™ Images Multi Focus Professional Edition (for PC only)
Get the most from your Motic™ microscope system with purposeful and thoughtfully designed software from Motic™. Motic™ Images Multi Focus Professional Edition (for PC only) captures and combines up to 100 images taken at different focusing levels to produce a single, fully focused image with great depth of field.
Brand:  Motic™ SE990794
Additional Details : Weight : 0.01000kg
Product Code. 15445222
  • £169.00 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
  • Combines up to 100 images taken at different focal depths.
  • The complex software algorithm will take into account image shift to create an in-focus image with great depth of field.
  • Available as a separate purchase or in the Motic™ Images Advanced package.
  • Windows™ XP or 2000 are recommended to run this software.
  • Compatible with Motic™ Images Plus 2.0 and integrated into Motic™ Advanced 3.2.
Specifications
Specifications
Software
Image stitching software
Windows™ XP or 2000
Microscope/camera/computer