Learn More
Motic™ Images Assembly Professional Edition (for PC only)
Click to view available options
For Use With (Equipment):
Microscope/camera/computer
Unit Size:
Each
Description
- Motic™ Images Assembly Pro will combine up to 400 images (20X20) images taken in one plane.
- The complex software algorithm will take into account overlap and shifting to produce high-magnification images with a large field of view.
- Motic™ Images Assembly Pro is contained within the Motic™ Images Advanced package and is also available as a separate purchase.
- Windows™ XP or 2000 are recommended to run this software.
- Compatible with Motic™ Images Plus 2.0 and integrated into Motic™ Advanced 3.2
Specifications
Specifications
| Type | Software |
| Description | Image stitching software |
| For Use With (Application) | Windows™ XP or 2000 |
| For Use With (Equipment) | Microscope/camera/computer |