missing translation for 'onlineSavingsMsg'
Learn More

Cytiva IMAC Sepharose™ 6 Fast Flow Affinity Media

Product Code. 10372810 Shop All Cytiva Products
Click to view available options
Quantity:
25 mL
100 mL
Particle Size:
45 to 165 μm
90 μm
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 10372810

Brand: Cytiva 17092108

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for purifying proteins and peptides that have an affinity for metal ions. Cytiva IMAC Sepharose 6 Fast Flow is composed of cross-linked 6% agarose beads modified with a novel chelating ligand immobilized to the base matrix.

  • Uncharged IMAC resin allows possibility to charge with metal ion of choice for optimized selectivity
  • Used for ions such as histidine-tagged proteins but also untagged recombinant or native proteins
  • High protein binding capacity
  • Coupling chemistry and novel ligand designed to give minimal ligand leakage
  • Available HiTrap™ and HiPrep™ pre-packed formats for easy scale-up and process development

 

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Stability Stable in commonly used aqueous Buffers: 0.01 M HCl, 1 M NaOH, 70% Acetic Acid, 2% SDS, 30% 2-Propanol
For Use With (Application) For Purifying Proteins and Peptides That Have an Affinity for Metal Ions
Storage Requirements 4°C to 30°C
Product Type IMAC Sepharose 6 Fast Flow Media
Flow Rate 250 to 400 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.