missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ILVBL Monoclonal Antibody (OTI8A12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525585
This item is not returnable.
View return policy
Description
ILVBL Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The protein encoded by this gene shares similarity with several thiamine pyrophosphate-binding proteins identified in bacteria, yeast, and plants. The highest degree of similarity is found with bacterial acetolactate synthases (AHAS), which are enzymes that catalyze the first step in branched-chain amino acid biosynthesis.
Specifications
| ILVBL | |
| Monoclonal | |
| 0.56 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A1L0T0 | |
| ILVBL | |
| Full length human recombinant protein of ILVBL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Monkey | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI8A12 | |
| Unconjugated | |
| ILVBL | |
| 209L8; 2-hydroxyacyl-CoA lyase 1 like; 2-hydroxyacyl-CoA lyase 2; acetolactate synthase-like protein; 5830463I21; acetolactate synthase homolog; acetolactate synthase-like protein; AHAS; AI415009; HACL1L; ILV2H; ilvB (bacterial acetolactate synthase)-like; ilvB acetolactate synthase like; ILVBL; ilvB-like protein | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10994, 362843 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction