missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILVBL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £316.00
Specifications
| Antigen | ILVBL |
|---|---|
| Concentration | 0.20 mg/mL |
| Dilution | Western Blot 1:1000, ICC/IF (Negative) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18238305
|
Novus Biologicals
NBP1-78433SS |
0.025 mL |
£150.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18230596
|
Novus Biologicals
NBP1-78433 |
0.1 mL |
£316.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILVBL Polyclonal specifically detects ILVBL in Human samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
| ILVBL | |
| Western Blot 1:1000, ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Epigenetics | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535 | |
| ILVBL | |
| IgG | |
| Affinity Purified |
| 0.20 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human | |
| A1L0T0 | |
| 10994 | |
| A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title