missing translation for 'onlineSavingsMsg'
Learn More

ILVBL Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18429954
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429954 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429954 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP178433JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ILVBL Polyclonal antibody specifically detects ILVBL in Human samples. It is validated for Western Blot, ICC/IF (Negative)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILVBL
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ICC/IF (Negative)
Formulation 50mM Sodium Borate
Gene Alias acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 10994
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.