missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILVBL Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78433G
This item is not returnable.
View return policy
Description
ILVBL Polyclonal specifically detects ILVBL in Human samples. It is validated for Western Blot.
Specifications
| ILVBL | |
| Polyclonal | |
| Western Blot 1:1000 | |
| A1L0T0 | |
| ILVBL | |
| A synthetic peptide made to an internal portion of the human ILVBL protein (between residues 350-500) [UniProt A1L0T0] | |
| 0.1 mL | |
| Epigenetics | |
| 10994 | |
| Store at 4C in the dark. |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| acetolactate synthase homolog, acetolactate synthase-like protein, AHASFLJ39061,209L8, EC 2.2.1, EC 2.2.1.-, ILV2H, ilvB (bacterial acetolactate synthase)-like, IlvB-like protein, MGC1269, MGC19535 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction