missing translation for 'onlineSavingsMsg'
Learn More

ILT7/CD85g/LILRA4 Antibody (17G10.2), DyLight 488, Novus Biologicals™

Product Code. 18407727 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407727 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407727 Supplier Novus Biologicals Supplier No. NBP262226G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT7/CD85g/LILRA4 Monoclonal antibody specifically detects ILT7/CD85g/LILRA4 in Human samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILT7/CD85g/LILRA4
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 17G10.2
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member G, CD85g, CD85g antigen, ILT-7, ILT7MGC129598, Immunoglobulin-like transcript 7, leukocyte immunoglobulin-like receptor subfamily A member 4, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 4, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member4, MGC129597
Host Species Mouse
Immunogen CD85g/ ILT7, a member of leukocyte immunoglobulin-like receptor family expressed on plasmacytoid dendritic cells, but not on myeloid dendritic cells and other peripheral blood leukocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.