missing translation for 'onlineSavingsMsg'
Learn More

ILT6/CD85e/LILRA3 Antibody (060), DyLight 550, Novus Biologicals™

Product Code. 18353801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353801 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353801 Supplier Novus Biologicals Supplier No. NBP290228R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ILT6/CD85e/LILRA3 Monoclonal specifically detects ILT6/CD85e/LILRA3 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILT6/CD85e/LILRA3
Applications Flow Cytometry
Classification Monoclonal
Clone 60
Conjugate DyLight 550
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member E, CD85e antigen, HM31, ILT-6, ILT6CD85e, Immunoglobulin-like transcript 6, leucocyte immunoglobulin-like receptor, Leukocyte immunoglobulin-like receptor 4, leukocyte immunoglobulin-like receptor A3, leukocyte immunoglobulin-like receptor subfamily A member 3, leukocyte immunoglobulin-like receptor, subfamily A (without TM domain), member3, LIR4CD85E, LIR-4HM43e3, Monocyte inhibitory receptor HM43/HM31
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ILT6/CD85e/LILRA3 (AAH28208.1; Met1-Glu439).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11026
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.