missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT5/CD85a/LILRB3 Antibody (MM0413-9S32) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £466.00
Specifications
| Antigen | ILT5/CD85a/LILRB3 |
|---|---|
| Clone | MM0413-9S32 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18085007
|
Novus Biologicals
NBP2-11729-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18086764
|
Novus Biologicals
NBP2-11729 |
0.1 mg |
£466.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ILT5/CD85a/LILRB3 Monoclonal specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| ILT5/CD85a/LILRB3 | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
| LILRB3 | |
| Protein G purified |
| MM0413-9S32 | |
| Monoclonal | |
| Purified | |
| RUO | |
| AAB68668 | |
| 11025 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title