missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT5/CD85a/LILRB3 Antibody (222821), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1806I
This item is not returnable.
View return policy
Description
ILT5/CD85a/LILRB3 Monoclonal antibody specifically detects ILT5/CD85a/LILRB3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| ILT5/CD85a/LILRB3 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11025 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| 222821 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| CD85 antigen-like family member A, CD85a antigen, HL9Immunoglobulin-like transcript 5, ILT-5, ILT5CD85a, Leukocyte immunoglobulin-like receptor 3, leukocyte immunoglobulin-like receptor subfamily B member 3, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR3CD85A, LIR-3MGC138403, member 3, Monocyte inhibitory receptor HL9, PIRB | |
| Mouse myeloma cell line NS0-derived recombinant human LILRB3/CD85a/ILT5, Gly24-Glu443, Accession # AAB68668 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction