missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ILT3/CD85k/LILRB4 Antibody (U.Texas patent anti-LILRB4) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28106-1mg
This item is not returnable.
View return policy
Description
ILT3/CD85k/LILRB4 Monoclonal antibody specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ILT3/CD85k/LILRB4 | |
| Monoclonal | |
| Unconjugated | |
| Q8NHJ6 | |
| Human | |
| Protein A purified | |
| RUO | |
| 11006 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| U.Texas patent anti-LILRB4 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD85 antigen-like family member K, CD85k, CD85k antigen, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5LILRB5, LIR-5subfamily B, member 4, member 4 | |
| LILRB4/ ILT3/ CD85k | |
| 1 mg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction