missing translation for 'onlineSavingsMsg'
Learn More

ILT3/CD85k/LILRB4 Antibody (1057706) [DyLight 650], Novus Biologicals™

Product Code. 30733051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733051 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733051 Supplier Novus Biologicals Supplier No. FAB24252W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT3/CD85k/LILRB4 Monoclonal antibody specifically detects ILT3/CD85k/LILRB4 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILT3/CD85k/LILRB4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1057706
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CD85 antigen-like family member K, CD85k, CD85k antigen, ILT-3, ILT3CD85K, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR5LILRB5, LIR-5subfamily B, member 4, member 4
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived recombinant human LILRB4/CD85k/ILT3 protein, Pro17-His257, Accession # ABM86208
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11006
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.