missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILT11/LILRA5 Rabbit anti-Human, Clone: 18, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90323-50UL
This item is not returnable.
View return policy
Description
ILT11/LILRA5 Monoclonal specifically detects ILT11/LILRA5 in Human samples. It is validated for ELISA.
Specifications
| ILT11/LILRA5 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 353514 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 18 | |
| ELISA 1:5000-1:10000 | |
| CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7 | |
| Recombinant Human ILT11/LILRA5 Protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction