missing translation for 'onlineSavingsMsg'
Learn More

ILT1/CD85h/LILRA2 Antibody (4D7), Novus Biologicals™

Product Code. 18376079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376079 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376079 Supplier Novus Biologicals Supplier No. H00011027M14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ILT1/CD85h/LILRA2 Monoclonal antibody specifically detects ILT1/CD85h/LILRA2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ILT1/CD85h/LILRA2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4D7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH27916
Gene Alias CD85 antigen-like family member H, CD85h, CD85h antigen, ILT-1, ILT1CD85H, Leukocyte immunoglobulin-like receptor 7, leukocyte immunoglobulin-like receptor subfamily A member 2, leukocyte immunoglobulin-like receptor subfamily A member 2 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 2, LIR7LIR-7Immunoglobulin-like transcript 1
Host Species Mouse
Immunogen LILRA2 (AAH27916, 1 a.a. ~ 466 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTPILTVLICLGLSLGPRTHVQAGHLPKPTLWAEPGSVIIQGSPVTLRCQGSLQAEEYHLYRENKSASWVRRIQEPGKNGQFPIPSIAREHAGRYHCQYYSHNHSSEYSDPLELVVTGAYGKPTLSALPSPVVTLGGNVTLQCVSQVAFDGFILCKEGEDEHPQRLNSHSHARGWSWAIFSVGPVSPSRRWSYRCYAYDSNSPYVWSLPSDLLELLVPGVSKKPSLSVQPGPMVAPGESLTLQCVSDVGYDRFVLYKEGERDFLQRPGWQPQAGLSQANFTLGPVSPSHGGQYRCYSAHNLSSEWSAPSDPLDILITGQFYDRPSLSVQPVPTVAPGKNVTLLCQSRGQFHTFLLTKEGAGHPPLHLRSEHQAQQNQAEFRMGPVTSAHVGTYRCYSSLSSNPYLLSLPSDPLELVVSASLGQHPQDYTVENLIRMGVAGLVLVVLGILLFEAQHSQRSLQDAAGR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11027
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.