missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILP-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567914
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family including IAP, XIAP/ILP-1/MIHA, and Livin/KIAP that bind to and inhibits specific proteases. A novel member in the IAP protein family was recently identified and designated ILP-2 for IAP-like protein-2. ILP-2 has high homology to ILP-1, but is encoded by a distinct gene that is solely expressed in testis of tested normal human tissues. ILP-2, unlike ILP-1, has no inhibitory effect on Fas and TNF induced apoptosis, but potently inhibits apoptosis induced by overexpression of Bax or by coexpression of caspase-9 with Apaf-1. ILP-2 interacts with the processed caspase-9. These results suggest that ILP-2 is a novel IAP family member with restricted specificity for caspase-9.
Specifications
| ILP-2 | |
| Polyclonal | |
| Unconjugated | |
| BIRC8 | |
| ILP-2, ILP2, hILP2; IAP-like protein 2; baculoviral IAP repeat-containing 8; baculoviral IAP repeat-containing protein 8; inhibitor of apoptosis-like protein 2; testis-specific inhibitor of apoptosis | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 112401 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96P09 | |
| BIRC8 | |
| Synthesized peptide derived from human ILP-2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction