missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ILP-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14276
This item is not returnable.
View return policy
Description
PA1-4276 detects the ILP-2 protein in human samples. PA1-4276 has been successfully used in Western blot procedures. PA1-4276 immunogen corresponds to amino acid residues 71-84 of the human ILP-2 protein.
Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family including IAP, XIAP/ILP-1/MIHA, and Livin/KIAP that bind to and inhibits specific proteases. A novel member in the IAP protein family was recently identified and designated ILP-2 for IAP-like protein-2. ILP-2 has high homology to ILP-1, but is encoded by a distinct gene that is solely expressed in testis of tested normal human tissues. ILP-2, unlike ILP-1, has no inhibitory effect on Fas and TNF induced apoptosis, but potently inhibits apoptosis induced by overexpression of Bax or by coexpression of caspase-9 with Apaf-1. ILP-2 interacts with the processed caspase-9. These results suggest that ILP-2 is a novel IAP family member with restricted specificity for caspase-9.
Specifications
| ILP-2 | |
| Polyclonal | |
| Unconjugated | |
| BIRC8 | |
| baculoviral IAP repeat containing 8; baculoviral IAP repeat-containing 8; baculoviral IAP repeat-containing protein 8; BIRC8; hILP2; IAP-like protein 2; ILP2; ILP-2; inhibitor of apoptosis-like protein 2; testis-specific inhibitor of apoptosis | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 112401 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q96P09 | |
| BIRC8 | |
| Synthetic peptide: E(71) E K G H E Y I N N I H L T(84). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction