missing translation for 'onlineSavingsMsg'
Learn More

Iloprost, MedChemExpress

Product Code. 16453776
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453776

Brand: Medchem Express HYA0096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Iloprost (ZK 36374; Ciloprost) is a prostacyclin (PGI2) analogue, involves in embryo development and inflammation improvement, and inhibits tumor metastasis. Iloprost can be used for peripheral vascular research.

Iloprost (ZK 36374

Specifications

Chemical Name or Material Iloprost
Physical Form Oil
CAS 78919-13-8
Color Colorless
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C22H32O4
Synonym Ciloprost ZK 36374
Solubility Information DMSO : 100 mg/mL (277.40 mM; Need ultrasonic)
SMILES O=C(O)CCC/C=C1C[C@@]2([H])C[C@@H](O)[C@H](/C=C/[C@@H](O)C(C)CC#CC)[C@@]2([H])C\1
Molecular Weight (g/mol) 360.49
Formula Weight 360.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.