missing translation for 'onlineSavingsMsg'
Learn More

Iloperidone, MedChemExpress

Product Code. 16497286
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497286 100 mg 100mg
16457328 10 mg 10mg
16403347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497286 Supplier Medchem Express Supplier No. HY17410
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Iloperidone (HP 873) is a D2/5-HT2 receptor antagonist. Iloperidone is an atypical antipsychotic for the schizophrenia symptoms.

Iloperidone (HP 873) is a D2/5-HT2 receptor antagonist. Iloperidone is an atypical antipsychotic for the schizophrenia symptoms.

Specifications

Chemical Name or Material Iloperidone
CAS 133454-47-4
Color White
Purity Grade Notes Research
Molecular Formula C24H27FN2O4
Quantity 100 mg
Synonym HP 873
Solubility Information DMSO : 50 mg/mL (117.24 mM; Need ultrasonic)
SMILES CC(C1=CC=C(OCCCN2CCC(C3=NOC4=C3C=CC(F)=C4)CC2)C(OC)=C1)=O
Molecular Weight (g/mol) 426.48
Formula Weight 426.48
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.