missing translation for 'onlineSavingsMsg'
Learn More

Iloperidone-d3, TRC

Product Code. 30240804
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240804 2.5 mg 2.5mg
30240734 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240804 Supplier Toronto Research Chemicals Supplier No. TRCI2672032.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071168-82-5
Molecular Formula C24H24D3FN2O4
Molecular Weight (g/mol) 429.5
Synonym 1-[4-[3-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]propoxy]-3-methoxyphenyl]ethanone-D3, HP 873-d3, Zomaril-d3;
SMILES FC1=CC=C2C(ON=C2C3CCN(CCCOC4=C(OC)C=C(C(C([2H])([2H])[2H])=O)C=C4)CC3)=C1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C24H27FN2O4/c1-16(28)18-4-7-21(23(14-18)29-2)30-13-3-10-27-11-8-17(9-12-27)24-20-6-5-19(25)15-22(20)31-26-24/h4-7,14-15,17H,3,8-13H2,1-2H3/i1D3
Chemical Name or Material Iloperidone ethanone-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.