missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
- Buffer-free and ready-to-use solutions in multiple formats
- Functionally tested for long PCR and sequencing
Order Info
Custom formulation, testing, packaging, and labeling are available
Specifications
Specifications
| Format | Liquid |
| Product Type | DNA Polymerase Mix dNTP (20mM each A,C,G,T) |
| Concentration | 20 mM |
| Purity | >99% |
| Quantity | 4 x 10 μm |
| Color | Colorless |
| For Use With (Application) | Functionally tested for long PCR and sequencing, Nucleic Acid amplification, labelling, mutagenesis, cDNA synthesis and expression profiling |
Safety and Handling
Product Identifier
- dNTP Mix - A C G T, 4 x 10
- EUH210-Safety data sheet available on request.
- UNKNOWN ACUTE-4,8 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 4,8% of components with unknown hazards to the aquatic environment.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?