missing translation for 'onlineSavingsMsg'
Learn More

Cytiva PlasmidPrep Midi Flow Kit

Product Code. 10176174 Shop All Cytiva Products
Click to view available options
No. of Reactions:
100
Unit Size:
100 reactions
This item is not returnable. View return policy

Product Code. 10176174

Brand: Cytiva 28904268

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

illustra plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA

illustra plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA. The kit allows for the purification of large yields of plasmid DNA that are proportional to the input sample volume. illustra plasmidPrep Midi Flow Kit can be used to process bacterial sample volumes of 25 to 50mL for high copy number plasmid DNA and up to 500mL for low copy number plasmid DNA. The kit consists of a high-performance, anion exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA. The kit contains fast-flow plasmid 250 columns, resuspension, lysis, neutralisation, wash, elution, and TE buffers, and a full protocol booklet with a detachable, quick reference protocol card.
  • High column capacity allows for a proportional yield for input sample volume of 25 to 50mL (for high copy number plasmid) and up to 500mL for low copy number plasmid.
  • Provides plasmid DNA with excellent purity and low endotoxin levels.
  • Increased ease of use due to minimal pipetting volume changes, color-coded kit components, and pre-equilibrated columns.
  • Modified alkaline lysis procedure.
  • Anion exchange chromatography.
  • Purification format is single-use, gravity flow illustra Fast-Flow plasmid 250 columns.
  • Greater than 70% (strain-dependent) of supercoiled pDNA.
  • Less than10 endotoxin units (EU) per ?g of plasmid DNA.
  • Kit consistently produces high yields of pure plasmid DNA with low levels of endotoxin contamination.
TRUSTED_SUSTAINABILITY

Specifications

Description illustra plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA. The kit allows for the purification of large yields of plasmid DNA that are proportional to the input sample volume.
For Use With (Application) Used For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification
No. of Reactions 100
Product Type illustra plasmidPrep Midi Flow Kit
Purity 2.5
Sample Volume 150 mL Culture
Test Time Approx.215 min
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.