Learn More
Cytiva MicroSpin™ S-300 HR Columns
For rapid purification of PCR products (>100 bp) from unincorporated primers (<20-mers) and nucleotides in a volume of 25 to 50μL using spin-column chromatography
Brand: Cytiva 27-5130-01
195.56 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : GH
Additional Details : CAS Number : 9048-46-8 Weight : 0.18000kg
Description
- Useful for purification of alkaline-denatured plasmid DNA prior to sequencing
- Requires <4 min. from sample application to collection of purified product
- Prepacked with Sephacryl™ S-300 HR media pre-equilibrated in TE buffer
- Percent sample recovery: >80%
- Tested in nickase, single- and double-stranded exonuclease and RNase assays
Specifications
Spin Column | |
Rapid purification of PCR products (>100bp) from unincorporated primers (<20-mers) and Nucleotides in a volume of 25 to 50μL using spin-column chromatography | |
Column | |
Each |