missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GFX PCR DNA and Gel Band Purification Kit

Product Code. 11329941 Shop All Cytiva Products
Click to view available options
Quantity:
10 Pack
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11329941

Brand: Cytiva 28903466

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

illustra GFX PCR DNA and Gel Band Purification Kit is designed for the rapid purification and concentration of PCR products or DNA fragments

illustra GFX PCR DNA and Gel Band Purification Kit is designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50bp to 10kb. This kit can be used to purify DNA from reaction volumes up to 100μl or agarose gel slices up to 900mg. The kit combines a versatile chaotropic buffer with a glass fibre matrix supported in a spin column for the purification of DNA from both solution and agarose gel. Typical recoveries range from 60% to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution. DNA purity is exceptional; 99.5% of contaminants are removed. Illustra GFX PCR DNA and Gel Band Purification Kit contains the following components in sufficient quantities: GFX columns, collection tubes, color-coded bottles of capture buffer, wash buffer, and two elution buffers (Tris-HCl and sterile water), and an instruction booklet.
  • For the isolation and concentration of DNA fragments (50 bp to 40 kb) from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction enzyme digests.
  • Fast and easy-to-use method with less than 10 minutes hands-on time.
  • Flexible 10 to 50μL elution volume for different DNA concentration needs.
  • Improved column design with increased maximum volume, holding of up to 800μL liquid volume (950μL without a cap).
  • Highly pure DNA is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning.
  • Addition of a visual color indicator to the capture buffer ensures optimal pH for maximum DNA binding and recovery.
  • Two choices of elution buffer provided to fit downstream applications.
  • Color-coded caps for ease of use.
TRUSTED_SUSTAINABILITY

Specifications

Description Designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg.
Sample Type Agarose Gel Slices, DNA Solutions, Enzyme Reactions, PCR Mixtures
For Use With (Application) For the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb
Product Type illustra GFX PCR DNA and Gel Band Purification Kit, 10 purifications
Quantity 10 Pack
Sample Volume 50 bp-10 kb
Yield PCR 10 μl elution volume-65%, PCR 50 μl elution volume-82%, 300 mg agarose 10 μl elution volume-57%, 300 mg agarose 50 μl elution volume-91%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P270-Do not eat, drink or smoke when using this product.
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.