missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GenomiPhi HY DNA Amplification Kit

Product Code. 10247754 Shop All Cytiva Products
Click to view available options
No. of Reactions:
100 reactions
1000 reactions
25 reactions
Unit Size:
100 reactions
1000 reactions
25 reactions
This item is not returnable. View return policy

Product Code. 10247754

Brand: Cytiva 25660020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Sample Buffer, reaction buffer, enzyme mix, and control λ DNA

Highly efficient and representative whole-genome amplification yielding 40 to 50μg from nanogram amounts of DNA sample

  • Starting material can be purified DNA or nonpurified cell lysates
  • Simple protocol provides typical DNA of 40 to 50μg, average lengths >10kb
  • Less hands-on time compared to traditional isolation methods
  • Representative amplification of the whole genome
  • Automation-friendly protocol with great reproducibility
  • High-quality DNA for array CGH, high-throughput genotyping, DNA archival, PCR, restriction enzyme digestion, hybridization, and cloning
  • Contains all of the components necessary for genomic DNA preparation by isothermal strand displacement amplification

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the kit at -70°C.The enzyme mix must be stored at -70°C; all other components may be stored at -20°C.Thaw components on ice and maintain at 0°C to 4°C during handling.
Description Illustra GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 50 μg yield from nanogram amounts of DNA sample.
For Use With (Application) Genomic DNA Amplification
Includes 2.25mL Sample Buffer, 2.25mL Reaction Buffer, 250μL Enzyme mix, 20mL Control DNA (lambda)
No. of Reactions 100 reactions
Product Type illustra GenomiPhi HY DNA Amplification Kit
Size > 10 kb
Test Time 4 h
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.